missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD298 Mouse, anti-Human, APC, Clone: LNH-94, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60159AZ
This item is not returnable.
View return policy
Description
- The LNH-94 antibody reacts with human CD298, the β3 subunit of the Na+/K+ ATPase
- The Na+/K+ ATPase is a 42 kDa type II transmembrane protein composed of four catalytic α subunits and three smaller glycosylated non-catalytic β subunits
- The Na+/K+ ATPase is important for establishing electrochemical gradients and maintaining normal resting membrane potential by coupling ATP hydrolysis to the active transport of Na+ and K+
- The β subunits have been reported to act as chaperones that stabilize the α subunits and occlude K+
- CD298 has been used to identify metastatic human cancer cells in mouse xenograft models and has been reported to modulate aspects of immunity, including HIV production, and T cell and B cell activation
- CD298 is found on leukocytes, platelets, and most other tissue types
Specifications
| CD298 | |
| Monoclonal | |
| APC | |
| 100 Tests | |
| Human | |
| B Cells, Granulocytes and Subsets, Lymphocytes, Mammary Cells, Monocytes, T Cells |
| Flow Cytometry | |
| LNH-94 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction