missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD29 (Integrin β1 chain) Mouse anti-Human, Brilliant Violet 510, Clone: MAR4, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747747
This item is not returnable.
View return policy
Description
CD29 (Integrin β1 chain) Monoclonal antibody specifically detects CD29 (Integrin β1 chain) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD29 (Integrin β1 chain) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| CD29 (Integrin β1 chain) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| MAR4 | |
| Brilliant Violet 510 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction