missing translation for 'onlineSavingsMsg'
Learn More

CD29 (Integrin β-1) Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments with Epredia™ CD29 (Integrin β-1) Ab-1.

£119.00 - £366.18

Specifications

Antigen CD29 (Integrin beta-1) Ab-1
Clone 4B7R
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Concentration Quantity Price Quantity & Availability  
Product Code Brand Concentration Quantity Price Quantity & Availability  
12641707
View Documents Promotion Details
Epredia
MS-596-P0
200μg/mL 100 μL
£119.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12661707
View Documents Promotion Details
Epredia
MS-596-P1
200μg/mL 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12631707
View Documents Promotion Details
Epredia
MS-596-P
200μg/mL 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651707
View Documents Promotion Details
Epredia
MS-596-P1ABX
1mg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12671707
View Documents Promotion Details
Epredia
MS-596-PABX
1mg/mL 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

CD29 (integrin Beta-1/gpIIa) is a cell adhesion molecule appearing on platelets, as the common Beta subunit of the very late activation antigen (VLA), and as a component of various protein complexes binding to extracellular matrix proteins. CD29 is expressed at the cell surface exclusively as part of a heterodimer, in association with one of at least nine different integrin alpha subunits (alpha1, alpha2, alpha3, alpha4, alpha5, alpha6, alpha7, alpha8, and alphav). With the exception of red blood cells and possible weak expression on granulocytes, CD29 is expressed on nearly all cell and tissue types.

Host Species: Mouse

Clone: 4B7R

Isotype: IgG1/κ

Species Reactivity: Human and Pig. Others not tested.

Epitope: Not determined

Immunogen: Human melanoma V+B2 cell line

Molecular Weight: 110kDa (nonreducing) and 130 (reducing)

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD29 (Integrin beta-1) Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Cell Adhesion
Human melanoma VplusB2 cell line
Primary
4B7R
Monoclonal
Concentrated
RUO
Human, Porcine
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications