Learn More
CD29 Hamster anti-Rat, FITC, Clone: HA2/5, BD
Hamster Monoclonal Antibody
£103.58 - £702.80
Specifications
| Antigen | CD29 |
|---|---|
| Clone | HA2/5 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15811349
|
BD Biosciences
561796 |
50 μg |
£107.00 £103.58 / Each Save £3.42 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15867948
|
BD Biosciences
555005 |
0.5 mg |
£720.00 £702.80 / Each Save £17.20 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Ha2/5 monoclonal antibody specifically binds to the 130 kDa integrin β1 chain (CD29). CD29 is expressed on the cell surface as a heterodimer with one of the distinct integrin α chains. With α1 through α6 (CD49a through CD49f), it forms the VLA-1 through VLA-6 complexes, respectively, and with αV (CD51), it forms αVβ
Specifications
| CD29 | |
| 0.5mg/mL | |
| Monoclonal | |
| Hamster | |
| Rat | |
| Rat glomerular epithelial cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| HA2/5 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgM κ | |
| Affinity Purified | |
| Itgb1; Integrin β1 chain; Integrin beta-1; VLA-4 subunit beta |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.