missing translation for 'onlineSavingsMsg'
Learn More

CD289 (TLR9) Monoclonal Antibody (eB72-1665), eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9099-95

 View more versions of this product

Product Code. 17170712

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

eB72-1665 is generated against a portion of human toll-like receptor 9 (aa 273-288), a molecule reported to be expressed predominantly intracellularly. TLR9 is a ∽115-120 kDa molecule, which mediates response to unmethylated CpG dinucleotides in bacterial DNA. CpG DNA induces a strong T-helper-1-like inflammatory response and the proliferation of TLR9-positive human B cells. When stimulated with CpG DNA, TLR9-deficient (TLR9-/-) mice lacked splenocyte proliferation, inflammatory cytokine production from macrophages, and dendritic cell maturation, as compared with normal mice. To date, at least twelve members of the Toll family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Members of the TLR family are involved in recognition and response to different microbial components including lipoproteins, peptidoglycans, and nucleic acids and play important roles in innate immunity and inflammation.

CD289 (TLR9) Monoclonal antibody specifically detects CD289 (TLR9) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD289 (TLR9)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9NR96
Rat
10 mg
Primary
Human
Antibody
IgG2a κ
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
eB72-1665
Unconjugated
Tlr9
CD289; Tlr9; toll like receptor 9; toll-like receptor 9; UNQ5798/PRO19605
Affinity chromatography
RUO
54106
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.