missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD284/MD-2 Complex Rat anti-Mouse, BV650, Clone: MTS510, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 740615
This item is not returnable.
View return policy
Description
CD284/MD-2 Complex Monoclonal antibody specifically detects CD284/MD-2 Complex in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD284/MD-2 Complex | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| MTS510 | |
| Brilliant Violet 650 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction