missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD281 (TLR1) Rat anti-Mouse, Oligo, Clone: TR23, BD™ AbSeq ™
Rat Monoclonal Antibody
Brand: BD Biosciences 940507
This item is not returnable.
View return policy
Description
CD281 (TLR1) Monoclonal antibody specifically detects CD281 (TLR1) in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD281 (TLR1) | |
| Monoclonal | |
| Oligo | |
| Cd281; Tlr1; Toll-like receptor 1; TIL; Toll/interleukin-1 receptor-like | |
| Mouse TLR1 Transfected Cell Line | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Single Cell 3' Sequencing | |
| Clone TR23 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction