missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Mouse anti-Rat, PE, Clone: JJ319, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A16199
This item is not returnable.
View return policy
Description
Specifications
CD28 | |
Monoclonal | |
PE | |
CD28RNA | |
Rat CD28 | |
RUO | |
25660 | |
Store at 2 - 8 °C. | |
IgG1 |
Flow Cytometry | |
JJ319 | |
proprietary buffer with 0.09% sodium azide | |
Mouse | |
50 μg | |
Primary | |
Rat | |
Liquid |