missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Monoclonal Antibody (10F3), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen CD2801
Code : 50
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry.Specifications
CD28 | |
Monoclonal | |
FITC | |
Cd28 | |
antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 | |
Mouse | |
purified | |
ASR | |
940 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
10F3 | |
PBS with BSA and 0.1% sodium azide | |
P10747 | |
Cd28 | |
Human CD28 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |