Learn More
CD28 Monoclonal Antibody (10F3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen CD28004
Code : 50
Additional Details : Weight : 0.50000kg
Description
CD28 antigen is a 44kDa disulfide linked homodimeric T cell specific surface glycoprotein. It is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes (approximately 95% of CD4+ cells and 50% of CD8+ cells). Mature thymocytes exhibit higher levels of CD28 than the immature cells. Activation of T cells results in enhanced CD28 expression. T cell activation requires two combined signals provided by antigen presenting cells.
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD28 | |
Monoclonal | |
Unconjugated | |
Cd28 | |
antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 | |
Mouse | |
purified | |
RUO, ASR/IVD | |
940 | |
4° C, store in dark | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
10F3 | |
PBS with BSA and 0.1% sodium azide | |
P10747 | |
Cd28 | |
Human CD28 | |
2 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |