missing translation for 'onlineSavingsMsg'
Learn More

CD28 Antibody (D665), DyLight 405, Novus Biologicals™

Product Code. 18383761 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383761 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383761 Supplier Novus Biologicals Supplier No. NBP281054V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD28 Monoclonal specifically detects CD28 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone D665
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Gene Symbols CD28
Host Species Rabbit
Immunogen The original D665 antibody was generated by immunizing mice alternately with A20 cells expressing mCD28 i.p. and with recombinant mCD28Ig s.c.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 940
Test Specificity This antibody binds to the laterally exposed C'D loop of CD28 and induces T cell proliferation without TCR ligation.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.