Learn More
CD279 (PD-1) Monoclonal Antibody (J116), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£7770.00 - £12950.00
Specifications
| Antigen | CD279 (PD-1) |
|---|---|
| Clone | J116 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
Description
The J116 MAb reacts with the human PD-1 (programmed death-1), a 55 kDa member of the immunoglobulin superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease. PD-1 is expressed predominantly on activated T and B lymphocytes. Two novel members of the B7 family have been identified as the PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these two PD-1 ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells. Binding of the J116 MAb inhibits PD-1 signal transduction, however, it does not block binding of the ligand PD-L1.
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow Cytometry, Functional AssaySpecifications
| CD279 (PD-1) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; Protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| J116 | |
| Flow Cytometry, Functional Assay | |
| Functional Grade | |
| Mouse | |
| Human | |
| Q15116 | |
| 5133 | |
| Primary | |
| 4° C | |
| Pdcd1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.