missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Hamster anti-Mouse , Clone: J43, BD Biosciences™
Hamster Monoclonal Antibody
Brand: BD Biosciences 566823
This item is not returnable.
View return policy
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Mouse samples. It is validated for Flow Cytometry, Blocking Assay
Specifications
| CD279 (PD-1) | |
| Monoclonal | |
| Aqueous buffered solution containing no preservative. | |
| Hamster | |
| 0.5 mg | |
| Primary | |
| Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions. |
| Flow Cytometry, Blocking Assay | |
| J43 | |
| Ly101; PD-1; Pdc1; Pdcd1; mPD-1; programmed cell death 1 protein; programmed cell death protein 1; protein PD-1 | |
| Syrian Hamster kidney cell line BKH transfected with Pdcd1 cDNA | |
| RUO | |
| Mouse | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction