missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD278 (ICOS) Monoclonal Antibody (ISA-3), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£154.00 - £340.00

Specifications

Antigen CD278 (ICOS)
Clone ISA-3
Concentration 0.6 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055850
View Documents
Invitrogen™
H060T03Y07-A
100 Tests
£340.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30054934
View Documents
Invitrogen™
H060T02Y07-A
25 Tests
£154.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD278 (ICOS) Monoclonal antibody specifically detects CD278 (ICOS) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD278 (ICOS)
0.6 μg/Test
Monoclonal
Mouse
Human
Q9Y6W8
29851
Primary
4°C, store in dark, DO NOT FREEZE!
ISA-3
Flow Cytometry
NovaFluor Yellow 730
RUO
PBS with 0.09% sodium azide; pH 7.2
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115
IgG1 κ
Affinity chromatography
ICOS
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.