missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD278 (ICOS) Monoclonal Antibody (ISA-3), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£154.00 - £340.00
Specifications
| Antigen | CD278 (ICOS) |
|---|---|
| Clone | ISA-3 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055695
|
Invitrogen™
H060T03R04-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054988
|
Invitrogen™
H060T02R04-A |
25 Tests |
£154.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD278 (ICOS) Monoclonal antibody specifically detects CD278 (ICOS) in Human samples. It is validated for Flow CytometrySpecifications
| CD278 (ICOS) | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q9Y6W8 | |
| 29851 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| ISA-3 | |
| Flow Cytometry | |
| NovaFluor Red 710 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
| IgG1 κ | |
| Affinity chromatography | |
| ICOS |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title