Learn More
Invitrogen™ CD278 (ICOS) Monoclonal Antibody (7E.17G9), PE, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£98.00 - £170.00
Specifications
| Antigen | CD278 (ICOS) |
|---|---|
| Clone | 7E.17G9 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15259369
|
Invitrogen™
12-9942-81 |
50 μg |
£98.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15269369
![]() |
Invitrogen™
12-9942-82 |
100 μg |
£170.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The 7E.17G9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells. Applications Reported: The 7E.17G9 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 7E.17G9 antibody has been tested by flow cytometric analysis of Con A-stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.Specifications
| CD278 (ICOS) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
| ICOS | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| ICOS |
| 7E.17G9 | |
| Flow Cytometry | |
| PE | |
| Rat | |
| Mouse | |
| Q9WVS0 | |
| 54167 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.