missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD275 (B7-H2) Monoclonal Antibody (987614)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561005
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.
Specifications
| CD275 (B7-H2) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9JHJ8 | |
| Icosl | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-H2, Met1-Lys279. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Inhibition Assays | |
| 987614 | |
| Unconjugated | |
| Icosl | |
| AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand | |
| Rat | |
| Protein A/G | |
| RUO | |
| 50723 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction