missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CD274(PD-L1)-PDCD1LG2(CD273)/CEN9q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | CD274 molecule |
Gene Alias | B7-H,B7H1,MGC142294,MGC142296,PD-L1,PDCD1L1,PDCD1LG1,PDL1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003375
|
Abnova™
FG0226.200UL |
200 μL |
£2030.00
200µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093365
|
Abnova™
FG0226.100UL |
100 μL |
£1235.00
100µL |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
B7-H,B7H1,MGC142294,MGC142296,PD-L1,PDCD1L1,PDCD1LG1,PDL1 | |
29126;80380 | |
Liquid |
Tube | |
CD274 molecule | |
CD274 | |
DNA/RNA |