missing translation for 'onlineSavingsMsg'
Learn More

CD274 Mouse anti-Human, PE, Clone: MIH1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 557924

 View more versions of this product

Product Code. 15819118

  • ONLINE EXCLUSIVE PRICE
    £217.00
    £207.22 / Each
    Save £9.78
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PD-L1 and PD-L2 are newly discovered members of the B7 family are the ligands for the program death 1 (PD-1) receptor. They are expressed on immature dendritic cells and mature dendritic cells. PD-L1, also called B7-H1, is expressed on antigen-presenting cells, including IFN- γ-stimulated monocytes, and activated human and murine dendritic cells. Monoclonal antibodies that block PD-L1 and PD-L2 on dendritic cells result in enhanced T cell proliferation and cytokine production. PD-L1 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, and on most carcinomas. Studies show overlapping functions of PD-L1 and PD-L2 and indicate an important role for the PD-L:PD-1 pathway in regulating T cell responses.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD274
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MIH1
B7-H1, PD-L1
Mouse
100 Tests
Immunology
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.