Learn More
Invitrogen™ CD270 (HVEM) Monoclonal Antibody (LH1), eBioscience™

Description
Description: The LH1 monoclonal antibody reacts with mouse Herpes Virus Entry Mediator (HVEM, TR2), a member of the TNF-receptor superfamily. HVEM is found on most cell types, including T cells, B cells, monocytes, neutrophils and dendritic cells. This receptor was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to this receptor protein has been shown to be part of the viral entry mechanism. The cytoplasmic region of HVEM was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. HVEM has also been demonstrated to be a unique ligand for BTLA (B and T lymphocyte attenuator). The conservation of the BTLA-HVEM interaction between mouse and human suggests that this system is an important pathway regulating lymphocyte activation and/or homeostasis in the immune response. The LH1 antibody has been reported as a blocking antibody, interfering with the HVEM-LIGHT interaction but not the HVEM-BTLA interaction. Applications Reported: This LH1 antibody has been reported for use in flow cytometric analysis and functional assays. Applications Tested: This LH1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined...
Specifications
Specifications
| Antigen | CD270 (HVEM) |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Clone | LH1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | TNFRSF14 |
| Gene Accession No. | Q80WM9 |
| Gene Alias | Atar; CD270; CD40-like protein; herpes virus entry mediator; herpes virus entry mediator A; Herpesvirus entry mediator A; HGNC:11912; HVEA; Hvem; LIGHTR; RP3-395M20.6; sCD2710; TNF receptor superfamily member 14; Tnfrs14; TNFRSF14; TR2; tumo; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.