missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | CD27/TNFRSF7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | LG.3A10 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 |
| Gene Symbols | CD27 |
| Host Species | Armenian Hamster |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?