missing translation for 'onlineSavingsMsg'
Learn More

CD27/TNFRSF7 Antibody (LG.3A10), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18397770 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397770 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397770 Supplier Novus Biologicals Supplier No. NBP281095AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD27/TNFRSF7
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone LG.3A10
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Gene Symbols CD27
Host Species Armenian Hamster
Immunogen This antibody was produced by injecting Armenian hamsters intraperitoneally with a mixture of two murine CD27/TNFRSF7 tag-expressing Armenian hamster fibroblast clones (AR0524 and AR0530) in Freund's adjuvant
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 939
Test Specificity This antibody is specific for mouse CD27/TNFRSF7, which, as in the human system, is largely restricted to cells of the T cell lineage, and is expressed on the great majority of both alpha-beta and gamma-delta T lymphocytes, and thymocytes.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.