missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD27 Recombinant Armenian Hamster Monoclonal Antibody (LG.3A10)

Armenian Hamster Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551910
This item is not returnable.
View return policy
Description
This antibody is specific for mouse CD27, which, as in the human system, is largely restricted to cells of the T cell lineage, and is expressed on the great majority of both alpha Beta and GammaDelta T lymphocytes, and thymocytes.
CD27 is a 50 kDa member of the tumor necrosis factor (TNF) receptor superfamily that includes CD40 and CD30. The TNF superfamily members are known for the regulation of cell proliferation and death. In contrast to the expression of other TNFR/TNF family members, expression of CD27 and its ligand CD70 is predominantly confined to lymphocytes. High expression levels of CD27 appear to be dependent on proper ligation of antigen receptors. CD70 expression requires additional co-stimulatory and/or pro-inflammatory signals. CD27 is expressed as a disulfide-linked homodimer on mature thymocytes, peripheral blood T cells and a subpopulation of B cells. Activation of T cells via TCR-CD3 complex results in upregulation of CD27 expression on the plasma membrane as well as in the release of its soluble 28-32 kDa form, sCD27, detected in the plasma, urine or spinal fluid. Soluble CD27 is an important prognostic marker of acute and chronic B cell malignancies. RgpA, a cystein proteinase, although activating T cells through the protease-activated receptors (PARs), degradates CD27 and counteracts T cell activation mediated by CD27 and its ligand CD70. CD27-binding protein (SIVA), a proapoptotic protein, can bind to this receptor and is thought to play an important role in the apoptosis induced by this receptor. Diseases associated with CD27 dysfunction include Lymphoproliferative Syndrome 2 and Autosomal Recessive Lymphoproliferative Syndrome.
Specifications
| CD27 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P26842, P41272 | |
| CD27 | |
| This antibody was produced by injecting Armenian hamsters intraperitoneally with a mixture of two murine CD27 tag-expressing Armenian hamster fibroblast clones (AR0524 and AR0530) in Freund's adjuvant. | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Immunoprecipitation, T-Cell Activation, Immunocytochemistry | |
| LG.3A10 | |
| Unconjugated | |
| CD27 | |
| CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
| Armenian Hamster | |
| Protein A | |
| RUO | |
| 21940, 500318, 939 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction