missing translation for 'onlineSavingsMsg'
Learn More

CD27 Mouse anti-Human , PE, Clone: O323, BD Biosciences™

Mouse Monoclonal Antibody

£56.50 - £221.70

Specifications

Antigen CD27
Clone O323
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17247734
View Documents
BD Biosciences
566945
25 Tests
ONLINE EXCLUSIVE PRICE
£73.85
£56.50 / 25 tests
Save £17.35
23% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17237734
View Documents
BD Biosciences
566944
100 Tests
ONLINE EXCLUSIVE PRICE
£231.00
£221.70 / 100 tests
Save £9.30
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD27
Flow Cytometry
PE
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Not Reported
Primary
O323
Monoclonal
Mouse
Human
CD27 antigen; TNFRSF7; S152; T14; Tp55
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.