missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Mouse anti-Human, BV711, Clone: M-T271, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 564893
This item is not returnable.
View return policy
Description
CD27 Monoclonal antibody specifically detects CD27 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications
| CD27 | |
| Monoclonal | |
| Brilliant Violet 711 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| Purified |
| Flow Cytometry | |
| M-T271 | |
| Vol. Per test: 5μL | |
| TNFRSF7; TNF receptor superfamily, member 7; T14; Tp55; S152 | |
| Human T-CLL cells | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV711 under optimum conditions and unconjugated antibody and free BD Horizon™ BV711 were removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction