missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: M-T271, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566450
This item is not returnable.
View return policy
Description
CD27 Monoclonal antibody specifically detects CD27 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.
Specifications
| CD27 | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 25 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
| Flow Cytometry | |
| M-T271 | |
| Mouse | |
| RUO | |
| Human, Baboon, Cynomolgus, Rhesus Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction