missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD27 Monoclonal Antibody (O323), Brilliant Ultra Violet™ 395, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£185.00 - £374.00
Specifications
| Antigen | CD27 |
|---|---|
| Clone | O323 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17816602
|
Invitrogen™
363-0279-41 |
25 Tests |
£185.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17866412
![]() |
Invitrogen™
363-0279-42 |
100 Tests |
£374.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD27 is a 50 kDa member of the tumor necrosis factor (TNF) receptor superfamily that includes CD40 and CD30. The TNF superfamily members are known for the regulation of cell proliferation and death. In contrast to the expression of other TNFR/TNF family members, expression of CD27 and its ligand CD70 is predominantly confined to lymphocytes. High expression levels of CD27 appear to be dependent on proper ligation of antigen receptors. CD70 expression requires additional co-stimulatory and/or pro-inflammatory signals. CD27 is expressed as a disulfide-linked homodimer on mature thymocytes, peripheral blood T cells and a subpopulation of B cells. Activation of T cells via TCR-CD3 complex results in upregulation of CD27 expression on the plasma membrane as well as in the release of its soluble 28-32 kDa form, sCD27, detected in the plasma, urine or spinal fluid. Soluble CD27 is an important prognostic marker of acute and chronic B cell malignancies. RgpA, a cystein proteinase, although activating T cells through the protease-activated receptors (PARs), degradates CD27 and counteracts T cell activation mediated by CD27 and its ligand CD70. CD27-binding protein (SIVA), a proapoptotic protein, can bind to this receptor and is thought to play an important role in the apoptosis induced by this receptor. Diseases associated with CD27 dysfunction include Lymphoproliferative Syndrome 2 and Autosomal Recessive Lymphoproliferative Syndrome.Specifications
| CD27 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
| CD27 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| O323 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 395 | |
| Mouse | |
| Human | |
| P26842 | |
| 939 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD27 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title