missing translation for 'onlineSavingsMsg'
Learn More

CD268 (BAFF Receptor), clone: 8A7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9117-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15298147

  • £62.63 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The 8A7 monoclonal antibody reacts with the human BAFF receptor, also known as B lymphocyte stimulator (BLyS protein) receptor, which is expressed on B cells. The ligand for this receptor, BAFF (B cell-activating factor of the TNF family) is a B cell survival factor and regulates CD21/35 and CD23 expression. Interaction of this ligand with its receptor causes elevated CD21/35 and CD23 expression, whereas receptor blockade has been shown to down-modulate expression.Applications Reported: The 8A7 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), and immunohistochemical staining. It has also been reported for use in functional assays. (Please use Functional Grade purified 8A7, cat. 16-9117, in functional assays.).Applications Tested: The 8A7 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD268 (BAFF Receptor)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q96RJ3
TNFRSF13C, BAFF-R, Tumor necrosis factor receptor superfamily member 13C, B-cell-activating factor receptor, BAFF-R, BLyS receptor 3, TNFRSF13C, BAFFR, BR3
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
8A7
Unconjugated
TNFRSF13C
Purified
TNFRSF13C
IgG2a, kappa
25μg
Store at 2-8°C.
Primary
115650
Videos

For Research Use Only.