missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD268 (BAFF Receptor) Mouse anti-Human, BUV661, Clone: 11C1, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 749942
This item is not returnable.
View return policy
Description
CD268 (BAFF Receptor) Monoclonal antibody specifically detects CD268 (BAFF Receptor) in Human samples. It is validated for Flow Cytometry
Specifications
| CD268 (BAFF Receptor) | |
| Monoclonal | |
| Brilliant Ultraviolet 661 | |
| BAFF Receptor; BAFF-R; BLyS receptor 3; BR3; TNFRSF13C | |
| Human BAFF-R Transfected Cell Line | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 11C1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction