missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD267 (TACI) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110998
This item is not returnable.
View return policy
Description
Immunogen sequence: ACFLKKRGDP CSCQPRSRPR QSPAKSSQDH AMEAGSPVST SPEPVETCSF CFPECRAPTQ ESAVTPGTPD PTCAGRWGCH TRTTVLQPCP HIPDSGLGIV CVPAQE Highest antigen sequence indentity to the following orthologs: Rat - 40%, Mouse - 43%.
Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.
Specifications
| CD267 (TACI) | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF13B | |
| 1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14836 | |
| TNFRSF13B | |
| Recombinant Protein corresonding to Human CD267 (TACI). Recombinant protein control fragment (Product #RP-102143). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction