missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD267 (TACI) Polyclonal Antibody

Product Code. 12868182
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12868182 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12868182 Supplier Invitrogen™ Supplier No. PA519972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is K562 cell lysate. PA5-19972 can be used with blocking peptide PEP-0097.

Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD267 (TACI)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TNFRSF13B
Gene Accession No. O14836
Gene Alias 1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B
Gene Symbols TNFRSF13B
Host Species Rabbit
Immunogen A synthetic peptide mapping at the amino terminus of human TACI.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23495
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.