Learn More
Invitrogen™ CD267 (TACI) Monoclonal Antibody (ebio8F10-3), APC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£91.50 - £167.00
Specifications
| Antigen | CD267 (TACI) |
|---|---|
| Clone | ebio8F10-3 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15380790
|
Invitrogen™
17-5942-81 |
50 μg |
£91.50
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15390790
![]() |
Invitrogen™
17-5942-82 |
100 μg |
£167.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The monoclonal antibody eBio8F10.3 recognizes TACI (Transmembrane Activator and Calcium-modulator and CAML Interactor) also known as CD267. TACI is a type III membrane protein belonging to the TNF receptor superfamily. The ligand for TACI has been shown to be BAFF (BLyS) and APRIL. TACI is expressed on the surface of a subset of maturing splenic B cells of the transitional type termed T2. Genetic studies have shown a role for TACI in both stimulation and inhibition of B cell proliferation. This actual role for TACI in the mouse has still not been determined. Recently human TACI has been shown to have repressive activities which inhibit BAFF-R function. Applications Reported: This ebio8F10-3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ebio8F10-3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.Specifications
| CD267 (TACI) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B | |
| TNFRSF13B | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| TNFRSF13B |
| ebio8F10-3 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Mouse | |
| Q9ET35 | |
| 57916 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.