Learn More
Invitrogen™ CD266 (TWEAK Receptor) Monoclonal Antibody (ITEM-1), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 14-9019-37
Description
The ITEM-1 mAb reacts with human TWEAK Receptor/Fn14 (fibroblast growth factor-inducible 14 kDa protein). Fn14 is distantly related to the TNFR family, containing one cysteine-rich domain in the extracellular region and a TNFR-associated factor binding domain but does not contain a death domain (DD) cytoplasmic region. Fn14 plays a role in TWEAK-induced endothelial cell migration, proliferation, and angiogenesis. TWEAK-induced cell death via Fn14 includes both apoptosis and necrosis and can be blocked by an anti-TWEAK antibody, CARL-1. Fn14 is expressed on HUVEC and in some cancer tissues but not on freshly isolated PBMCs. Fn14 mRNA expression has been identified during liver regeneration.
TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrate that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by binding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in the aorta and liver. It may also play a role in vascular cell migration and angiogenesis.
Spécification
| CD266 (TWEAK Receptor) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9NP84 | |
| TNFRSF12A | |
| Affinity chromatography | |
| RUO | |
| 51330 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
| ITEM-1 | |
| Unconjugated | |
| TNFRSF12A | |
| AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12A; TNFRSF12A; Tumor necrosis factor receptor superfamily member 12A; tumor necrosis factor receptor superfamily, member 12A; TweakR; TWEAK-R; Tweak-receptor; type I transmembrane protein Fn14 | |
| Mouse | |
| 2 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.