Learn More
CD266 (TWEAK Receptor) Monoclonal Antibody (ITEM-1), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9019-95
Description
The ITEM-1 mAb reacts with human TWEAK Receptor/Fn14 (fibroblast growth factor-inducible 14 kDa protein). Fn14 is distantly related to the TNFR family, containing one cysteine-rich domain in the extracellular region and a TNFR-associated factor binding domain but does not contain a death domain (DD) cytoplasmic region. Fn14 plays a role in TWEAK-induced endothelial cell migration, proliferation, and angiogenesis. TWEAK-induced cell death via Fn14 includes both apoptosis and necrosis and can be blocked by an anti-TWEAK antibody, CARL-1. Fn14 is expressed on HUVEC and in some cancer tissues but not on freshly isolated PBMCs. Fn14 mRNA expression has been identified during liver regeneration.
CD266 (TWEAK Receptor) Monoclonal antibody specifically detects CD266 (TWEAK Receptor) in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen)
Specifications
| CD266 (TWEAK Receptor) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9NP84 | |
| Mouse | |
| 10 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) | |
| ITEM-1 | |
| Unconjugated | |
| TNFRSF12A | |
| AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12A; Tnfrsf12a; Tumor necrosis factor receptor superfamily member 12A; tumor necrosis factor receptor superfamily, member 12A; TweakR; TWEAK-R; tweak-receptor; type I transmembrane protein Fn14 | |
| Affinity chromatography | |
| RUO | |
| 51330 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.