Learn More
CD265 (RANK) Monoclonal Antibody (R12-31), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£157.00 - £277.00
Specifications
Antigen | CD265 (RANK) |
---|---|
Clone | R12-31 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15207927
|
Affymetrix eBioscience
14-6612-82 |
100 μg |
£157.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15217927
|
Affymetrix eBioscience
14-6612-85 |
500 μg |
£277.00
500µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The R12-31 monoclonal antibody reacts with mouse RANK (Receptor activator of NF-kappaB), a recently cloned member of the TNFR superfamily with no significant homology to other members of this family. RANK ligand (RANKL/TRANCE/ OPGL) binds to RANK on dendritic cells, upregulates the expression of anti-apoptotic protein BcL-XL suggesting a role in dendritic cell survival. The cytoplasmic domain of RANK interacts with TRAF2, TRAF5 and TRAF6. Overexpression of RANK activates NF-kappaB and c-Jun-terminal kinase (JNK) pathways. Recent studies have shown that RANK interaction with TRAF6 activates NF-kappaB, whereas JNK activation is mediated through binding of RANK to TRAF2. Applications Reported: The R12-31 antibody has been reported for use in flow cytometric analysis. Applications Tested: The R12-31 antibody has been tested by flow cytometric analysis of mouse RANK transfected cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.Specifications
CD265 (RANK) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
TNFRSF11A | |
Primary | |
4° C | |
TNFRSF11A |
R12-31 | |
Flow Cytometry | |
Unconjugated | |
Rat | |
Mouse | |
O35305 | |
21934 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.