missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD264 (TRAIL-R4) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581077
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human TRAIL R4/CD264/TNFRSF10D.
TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. A fourth member of the emerging TRAIL receptor family, TRAIL-R4, has signaling capacity similar to that of TRAIL-R1 and TRAIL-R2 with respect to NF-kappaB activation, but differs in its inability to induce apoptosis. Like TRAIL-R1 and TRAIL-R2, this receptor shows widespread tissue expression. The human TRAIL-R4 gene has been mapped to chromosome 8p22-21, clustered with three other TRAIL receptors.
Specifications
| CD264 (TRAIL-R4) | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF10D | |
| CD264; DCR2; Decoy receptor 2; TNF receptor superfamily member 10d; TNF receptor-related receptor for TRAIL; TNF-related apoptosis-inducing ligand receptor 4; TNFRSF10D; TRAIL receptor 4; TRAIL receptor with a truncated death domain; TRAILR4; TRAIL-R4; TRUNDD; Tumor necrosis factor receptor superfamily member 10D; tumor necrosis factor receptor superfamily member 10d, decoy with truncated death domain; tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain; UNQ251/PRO288 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 8793 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 5 mg/mL | |
| PBS with no preservative | |
| Q9UBN6 | |
| TNFRSF10D | |
| Recombinant Human TRAIL R4/CD264/TNFRSF10D protein (Met1-His211). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido