missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD262 (TRAIL-R2) Hamster anti-Mouse , PE, Clone: MD5-1, BD Biosciences™
Hamster Monoclonal Antibody
Brand: BD Biosciences 566813
This item is not returnable.
View return policy
Description
CD262 (TRAIL-R2) Monoclonal antibody specifically detects CD262 (TRAIL-R2) in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD262 (TRAIL-R2) | |
| Monoclonal | |
| PE | |
| Tnfrsf10b; TRAILR2; DR5; TRICK2; KILLER; Ly98; MK | |
| Mouse DR5 Recombinant Protein | |
| RUO | |
| Mouse | |
| IgG κ |
| Flow Cytometry | |
| MD5-1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Hamster | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction