missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD26 Monoclonal Antibody (M-A261), FITC, eBioscience™
GREENER_CHOICE

Product Code. 15571427
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15571427 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15571427 Supplier Invitrogen™ Supplier No. BMS143FI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: CD26 is a 110 kD type II membrane protein encoded by the DPP4 gene. It is an antigenic enzyme expressed on the surface of most cell types and is associated with immune regulation, signal transduction and apoptosis. It also plays an integral role as a costimulatory molecule in T cell activation. CD26 may interact with extracellular matrix proteins such as fibronectin or collagen, CD45 and ADA. Immunohistochemistry: The antibody can be used to stain cryostat sections. Recommended for BMS143 is the alkaline phosphatase-anti-alkaline-phosphatase (APAAP), or peroxidase anti-peroxidase (PAP) procedures or the three stage immunoperoxidase technique on acetone-fixed cryostat sections. Flow cytometry: BMS143 is also suitable as primary antibody in staining for FACS analysis, BMS143FI can be used for direct staining of cells, BMS143BT can be used in a two-step procedure using the biotin-(strept-)avidin system. Recommended dilution is 1:20 as determined on A704 cells. Immunoprecipitation: BMS143 is also suitable for immunoprecipitation. Optimal dilutions should be determined by the individual laboratory for each application. Applications Tested: Flow Cytometry. Purity: >95%. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

CD26 (dipeptidyl peptidase IV, DPP IV), adenosine deaminase (ADA) binding protein) is a homodimeric atypical serine protease belonging to the prolyl oligopeptidase family. CD26 is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. CD26 identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. Further, CD26 is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. Alterations in CD26 peptidase activity are characteristic of malignant transformation, and the enzymatic activity increases dramatically with tumor grade and severity. CD26 is expressed in various blood cell types, but also in cells that are histogenetically related to activated fibroblasts. Alterations in CD26 density have been reported on circulating monocytes and CD4+ T cells during rheumatoid arthritis and systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD26
Applications Flow Cytometry
Classification Monoclonal
Clone M-A261
Concentration 10 μL/Test
Conjugate FITC
Formulation TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4
Gene DPP4
Gene Accession No. P27487
Gene Alias ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10
Gene Symbols DPP4
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1803
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.