Learn More
CD253 (TRAIL) Monoclonal Antibody (N2B2), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£94.00 - £211.00
Specifications
Antigen | CD253 (TRAIL) |
---|---|
Clone | N2B2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15558166
|
Affymetrix eBioscience
17-5951-82 |
100 μg |
£211.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15548166
|
Affymetrix eBioscience
17-5951-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The N2B2 monoclonal antibody reacts with mouse TNF-related apoptosis-inducing ligand (TRAIL), a member of the TNF superfamily. TRAIL is not detected on the surface of freshly isolated T, B, or NK cells, but can be induced preferentially on CD3- NK1.1+ NK cells after stimulation with IL-2 or IL-15. N2B2 inhibits IL-2- or IL-15-activated NK cell cytotoxicity against mouse fibrosarcoma L929 target cells. Applications Reported: This N2B2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This N2B2 antibody has been tested by flow cytometric analysis of mouse CD253 transfected cells and can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD253 (TRAIL) Monoclonal antibody specifically detects CD253 (TRAIL) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD253 (TRAIL) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P50592 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
N2B2 | |
Flow Cytometry | |
APC | |
Rat | |
Mouse | |
22035 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.