Learn More
CD253 Mouse anti-Human, PE, Clone: RIK-2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 550516
Description
The RIK-2 monoclonal antibody specifically binds to CD253 which is also known as TRAIL (TNF-Related Apoptosis-Inducing Ligand) and Apo2 ligand (APO-2L). CD253 is a member of the Tumor Necrosis Factor Superfamily and is encoded by the TNFSF10 gene. CD253 is a type II membrane protein that may be expressed as a full-length, cell surface associated protein as well as in a soluble form. Both surface and soluble forms of TRAIL rapidly induce apoptosis by a wide range of cell lines but not by normal cells. TRAIL is expressed by activated T cells, NK cells, dendritic cells, monocytes and by a variety of non-lymphoid cells. TRAIL can bind to and exert apoptosis through DR4 (TRAIL-R1) and DR5 (TRAIL-R2) receptors. It can also bind to decoy receptors, including DcR1/TRID/TRAIL-R3 and DcR2/TRUNDD/TRAIL-R4, and possibly OPG/TNFRSF11B, which may serve to regulate TRAIL activity. TRAIL has been shown to be involved in T cell-mediated cytotoxicity, however its mechanism of action remains to be fully elucidated. The RIK-2 clone was selected based on its ability to block TRAIL-mediated cytotoxic activity.
Specifications
| CD253 | |
| Monoclonal | |
| 0.2mg/mL | |
| TNFSF10; TRAIL; APO-2L; TL2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| RIK-2 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human TRAIL | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.