Learn More
Invitrogen™ CD25 Recombinant Mouse Monoclonal Antibody (AHT54)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551903
Description
AHT54 recognises a determinant close to the IL-2 binding site. Preparation: BALB/c mice were immunized with human T lymphoblasts and spleen cells of the immunized animals were fused with X63-Ag8.653 mouse myeloma cells. The supernatants of the resultant hybridomas were screened by a proliferation inhibition assay (inhibition of the human T lymphoblast response to IL-2). Two of those (the clones AHT54 and AHT107) that produced antibodies which inhibited IL-2 dependent proliferation of human T lymphoblasts were selected, cloned and recloned.
CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.
Specifications
| CD25 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P01589 | |
| IL2RA | |
| BALB/c mice were immunized with human T lymphoblasts and spleen cells of the immunized animals were fused with X63-Ag8.653 mouse myeloma cells. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Inhibition Assays | |
| AHT54 | |
| Unconjugated | |
| IL2RA | |
| ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
| Mouse | |
| Protein A | |
| RUO | |
| 3559 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.