missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Rat, PE, Clone: PC61.5, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB34-PE-100ug
Additional Details : Weight : 0.01000kg
Specifications
CD25 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Il2ra | |
Protein A/G Purified | |
RUO | |
16184 | |
IgG1 λ |
Flow Cytometry | |
PC61.5 | |
PE | |
NP_032393 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |