Learn More
CD25 Rat anti-Mouse, APC-Cy7, Clone: PC61, BD
Rat Monoclonal Antibody
£108.72 - £292.84
Specifications
| Antigen | CD25 |
|---|---|
| Clone | PC61 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15860709
|
BD Biosciences
561038 |
25 μg |
£114.00 £108.72 / Each Save £5.28 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15808978
|
BD Biosciences
557658 |
0.1 mg |
£305.00 £292.84 / Each Save £12.16 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PC61 antibody reacts with CD25, the low-affinity IL-2 Receptor α chain (IL-2Rα, p55) expressed on activated T and B lymphocytes from all mouse strains tested. IL-2Rα by itself is not a signaling receptor. However, it can combine with IL-2 Receptor β (CD122) and γc (CD132) chains to form high-affinity, signaling receptor complexes for IL-2. Resting T and B lymphocytes and resting and activated NK cells do not express IL-2Rα. CD25 is transiently expressed at a low level during normal B-cell development in the bone marrow on the CD45R/B220low TdT- sIg- Pre-B/Pre-B-II and CD45R/B220low TdT- sIgM+ sIgD- immature B stages, but not on the CD45R/B220low TdT+ sIg- Pro-B/Pre-B-I stage nor on CD45R/B220high TdT- sIgM+ sIgD+ mature B cells. It is expressed at a higher level during a very early stage of T-cell development in fetal and adult thymus. Peripheral CD25+CD4+ lymphocytes called regulatory T (Treg) cells are involved in the maintenance of self-tolerance. It has also been reported that dendritic cells express CD25, recognized by mAb 7D4 (Cat. No. 553068). The PC61 antibody recognizes an epitope of CD25 which is distinct from the IL-2 binding site and from those recognized by mAbs 3C7 (Cat. No. 557364) and 7D4 (Cat. No. 553068). It blocks binding of IL-2 to CD25, presumably by inducing a conformational change in CD25.
Specifications
| CD25 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| IL-2-dependent cytolytic mouse T-cell clone B6.1 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| PC61 | |
| Flow Cytometry | |
| APC-Cyanine7 | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 λ | |
| Affinity Purified | |
| Interleukin-2 receptor alpha chain; IL-2RA; IL-2Rα; Il2ra; IL-2R p55 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.