missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD25 Monoclonal Antibody (BC96), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£154.00 - £340.00
Specifications
| Antigen | CD25 |
|---|---|
| Clone | BC96 |
| Concentration | 0.2 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055625
|
Invitrogen™
H052T03Y06-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054926
|
Invitrogen™
H052T02Y06-A |
25 Tests |
£154.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD25 Monoclonal antibody specifically detects CD25 in Human samples. It is validated for Flow CytometrySpecifications
| CD25 | |
| 0.2 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01589 | |
| 3559 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| BC96 | |
| Flow Cytometry | |
| NovaFluor Yellow 700 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
| IgG1 κ | |
| Affinity chromatography | |
| IL2RA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title