missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/IL-2R alpha Antibody (OX-39), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64761AF532
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD25/IL-2R alpha | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IL2RA | |
| Stimulated Rat T cells | |
| Primary | |
| NB100-64761 recognizes the alpha chain of rat CD25, otherwise known as IL-2 receptor alpha, a 55kDa type I membrane glycoprotein, expressed by activated T cells but not resting lymphocytes. CD25 is also expressed by dendritic cells found in the thymus medulla. *This clone has been described reacting with paraffin-embedded material following PLP fixation (periodate-lysine-paraformaldehyde). Clone OX-39 has been shown to weakly inhibit the binding of IL-2 to Con-A stimulated spleen blasts. | |
| Store at 4°C in the dark. |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OX-39 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Mouse | |
| 0.1 mL | |
| 3559 | |
| Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction