missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/IL-2R alpha Antibody (IL2RA/4375R), Novus Biologicals™
Rabbit Monoclonal Antibody
£242.00 - £440.00
Specifications
| Antigen | CD25/IL-2R alpha |
|---|---|
| Clone | IL2RA/4375R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662974
|
Novus Biologicals
NBP3-20509-100ug |
100 μg |
£440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621754
|
Novus Biologicals
NBP3-20509-20ug |
20 μg |
£242.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD25/IL-2R alpha | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
| 10mM PBS with 0.05% BSA | |
| 3559 | |
| IgG κ | |
| Protein A or G purified |
| IL2RA/4375R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Recombinant fragment (around aa1-250) corresponding to human CD25/IL-2R alpha | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title