missing translation for 'onlineSavingsMsg'
Learn More

CD25/IL-2R alpha Antibody (Daclizumab) - Azide and BSA Free, Novus Biologicals™

Product Code. 18356001 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356001 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356001 Supplier Novus Biologicals Supplier No. NBP275901SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD25/IL-2R alpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Blocking Assay, Neutralization
Classification Monoclonal
Clone Daclizumab
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Block/Neutralize
Formulation PBS with 0.02% Proclin 300
Gene Alias biosimilar, biosimilars, CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Gene Symbols IL2RA
Host Species Human
Immunogen Cultured T-cells derived from a patient with Mycosis fungoides.
Purification Method Protein A purified
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 3559.0
Test Specificity Binds to p55 chain of IL2R on human T cells, Tac antigen, which is expressed on T cells activated by mitogens, soluble antigens, and alloantigens but not on resting T cells, thymocytes, B cells, monocytes, activated B cells, leukemic T cell blasts from patients with acute lymphoblastic leukemia, and long-term T cell lines.
Target Species Human, Cynomolgus Monkey, Rhesus Macaque
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.