missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | CD25/IL-2R alpha |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | BLR157J |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Formulation | Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA and 0.09% Sodium Azide |
| Gene Alias | CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR |
| Host Species | Rabbit |
| Immunogen | residues 22-213 (ECD) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?