Learn More
CD25 (IL-2 Receptor α) Mouse anti-Human, BUV563, Clone: 2A3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612919
This item is not returnable.
View return policy
Description
CD25 (IL-2 Receptor α) Monoclonal antibody specifically detects CD25 (IL-2 Receptor α) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD25 (IL-2 Receptor α) | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| 25 Tests | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 2A3 | |
| Mouse | |
| RUO | |
| CD25 (IL-2 Receptor α) | |
| Refrigerate (2 to 8°C) |