missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD249 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581379
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse ENPEP/Aminopeptidase A.
CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.
Specifications
| CD249 | |
| Polyclonal | |
| Unconjugated | |
| Enpep | |
| 6030431M22Rik; Aminopeptidase A; aminopeptidase-A; APA; AP-A; BP1; Bp-1/6C3; BP-1/6C3 antigen; CD249; differentiation antigen gp160; EAP; ENPEP; Glutamyl aminopeptidase; glutamyl aminopeptidase (aminopeptidase A); gp160; Ly51; Ly-51 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 13809 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| P16406 | |
| Enpep | |
| Recombinant Mouse ENPEP/Aminopeptidase A Protein (Arg41-Pro945). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction